5-Fluoro-o-anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Fluoro-2-anisaldehyde, 5-Fluoro-2-methoxybenzaldehyde

B24684-03 B24684-06 B24684-14
CAAAAB24684-03EA 31.76 CAD
CAAAAB24684-03 CAAAAB24684-06 CAAAAB24684-14
5-Fluoro-o-anisaldehyde 98%
5-Fluoro-o-anisaldehyde

Purity: 98% CAS No.: 19415-51-1 Molecular Formula: C8H7FO2 Formula Weight: 154.14 MDL No.: MFCD00143458

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143458
CAS Number: 19415-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR