3,5-Dichlorobenzoyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24676-06
B24676-14
B24676-22
CAAAAB24676-06EA
80.9
CAD
CAAAAB24676-06
CAAAAB24676-14
CAAAAB24676-22
3,5-Dichlorobenzoyl chloride 96%
3,5-Dichlorobenzoyl chloride
Purity: 96%
CAS No.: 2905-62-6
Molecular Formula: C7H3Cl3O
Formula Weight: 209.46
Flash Point: 137°(278°F)
Refractive Index: 1.5820
Melting Point: 24-28°
Boiling Point: 245°
Beilstein Reference: 1940681
UN No.: 3261
MDL No.: MFCD00009817
Formula:
C₇H₃Cl₃O Boiling Pt: 245 °C Melting Pt: 24…28 °C Density: 1.478 Flash Pt: 137 °C (278 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009817 CAS Number: 2905-62-6 EINECS: 220-813-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...