3-Bromo-p-toluidine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24670-06
B24670-14
CAAAB24670-06EA
70.31
CAD
CAAAB24670-06
CAAAB24670-14
3-Bromo-p-toluidine 97%
3-Bromo-p-toluidine
Purity: 97%
CAS No.: 7745-91-7
Molecular Formula: C7H8BrN
Formula Weight: 186.06
Flash Point:
Formula:
C₇H₈BrN Boiling Pt: 254…257 °C Melting Pt: 25…28 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00134176 CAS Number: 7745-91-7 EINECS: 231-807-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...