1,2-Diaminocyclohexane mixture of isomers 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,2-Diaminocyclohexane, 1,2-Cyclohexanediamine
B24657-0B
B24657-30
CAAAB24657-0BEA
169.06
CAD
CAAAB24657-0B
CAAAB24657-30
1,2-Diaminocyclohexane mixture of isomers 99%
1,2-Diaminocyclohexane
Purity: 99%
CAS No.: 694-83-7
Molecular Formula: C6H14N2
Formula Weight: 114.19
Flash Point: 75°(167°F)
Refractive Index: 1.4870
Odor: Amine-like
pH: 12
Boiling Point: 191°
Beilstein Reference: 506142
UN No.: 2735
MDL No.: MFCD00001491
Formula:
C₆H₁₄N₂ MW: 114.19 g/mol Boiling Pt: 191 °C Melting Pt: 41…44 °C Density: 0.94 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001491 CAS Number: 694-83-7 EINECS: 211-776-7 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...