4-Ethylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B24656-03 B24656-06 B24656-14
CAAAB24656-03EA 41.4 CAD
CAAAB24656-03 CAAAB24656-06 CAAAB24656-14
4-Ethylphenylboronic acid 97%

Purity: 97% CAS No.: 63139-21-9 Molecular Formula: C8H11BO2 Formula Weight: 149.99 Melting Point: 150-154° Beilstein Reference: 3030638

Formula: C₈H₁₁BO₂
Melting Pt: 150…154 °C
Density: 1.125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00859377
CAS Number: 63139-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR