Diallyl phthalate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phthalic acid diallyl ester
B24648-22
B24648-36
CAAAB24648-22EA
37.13
CAD
CAAAB24648-22
CAAAB24648-36
Diallyl phthalate 97%
Diallyl phthalate
Purity: 97%
CAS No.: 131-17-9
Molecular Formula: C14H14O4
Formula Weight: 246.27
Flash Point: 166°(330°F)
Refractive Index: 1.5190
Odor: Pungent, makes eyes water
Melting Point: -70°
Boiling Point: 160-163°/4mm
Beilstein Reference: 1880877
UN No.: 3082
MDL No.: MFCD00008646
Formula:
C₁₄H₁₄O₄ Boiling Pt: 160…163 °C/4 mm Melting Pt: –70 °C Density: 1.120 Flash Pt: 166 °C (330 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008646 CAS Number: 131-17-9 EINECS: 205-016-3 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...