tert-Pentylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Amylamine
B24639-14
B24639-22
CAAAB24639-14EA
117.87
CAD
CAAAB24639-14
CAAAB24639-22
tert-Pentylamine 98%
tert-Pentylamine
Purity: 98%
CAS No.: 594-39-8
Molecular Formula: CH3CH2C(CH3)2NH2
Formula Weight: 87.17
Flash Point: -1°(30°F)
Refractive Index: 1.3995
Odor: Amine-like
Boiling Point: 76-78°
UN No.: 1106
MDL No.: MFCD00008056
Formula:
C₅H₁₃N Boiling Pt: 76…78 °C Melting Pt: –105 °C Density: 0.746 Flash Pt: –1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008056 CAS Number: 594-39-8 EINECS: 209-839-9 UN: 1106 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...