Isobutyrophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isopropyl phenyl ketone, 2-Methyl-1-phenyl-1-propanone, Phenyl isopropyl ketone, α-Methylpropiophenone

B24628-14 B24628-22
CAAAB24628-14EA 77.89 CAD
CAAAB24628-14 CAAAB24628-22
Isobutyrophenone 97%
Isobutyrophenone

Purity: 97% CAS No.: 611-70-1 Molecular Formula: C10H12O Formula Weight: 148.21 Flash Point: 84°(184°F) Refractive Index: 1.5170 Melting Point: 1° Boiling Point: 217° MDL No.: MFCD00008917

Formula: (C₆H₅)C(O)CH(CH₃)₂
Boiling Pt: 217 °C
Melting Pt: 1 °C
Density: 0.988
Flash Pt: 84 °C(184 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008917
CAS Number: 611-70-1
EINECS: 210-275-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR