2,4-Dimethoxybenzonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24621-06
B24621-14
CAAAB24621-06EA
22.75
CAD
CAAAB24621-06
CAAAB24621-14
CAAAB24621-22
2,4-Dimethoxybenzonitrile 99%
2,4-Dimethoxybenzonitrile
Purity: 99%
CAS No.: 4107-65-7
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Melting Point: 92-95°
Beilstein Reference: 1950512
UN No.: 3439
MDL No.: MFCD00001786
Formula:
C₉H₉NO₂ Boiling Pt: 283 °C (1013 hPa) Melting Pt: 92…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001786 CAS Number: 4107-65-7 EINECS: 223-886-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...