2,6-Dimethylphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24613-03
B24613-06
B24613-14
CAAAB24613-03EA
63.36
CAD
CAAAB24613-03
CAAAB24613-06
CAAAB24613-14
2,6-Dimethylphenylboronic acid 97%
2,6-Dimethylphenylboronic acid
Purity: 97%
CAS No.: 100379-00-8
Molecular Formula: C8H11BO2
Formula Weight: 149.99
Melting Point: 120-126° dec.
MDL No.: MFCD01009693
Formula:
C₈H₁₁BO₂ Melting Pt: 120…126 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01009693 CAS Number: 100379-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...