α,α,α-Trifluoro-2-tolualdehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24591-06
B24591-14
B24591-22
CAAAB24591-06EA
62.88
CAD
CAAAB24591-06
CAAAB24591-14
CAAAB24591-22
α,α,α-Trifluoro-2-tolualdehyde 98%
α,α,α-Trifluoro-2-tolualdehyde
Purity: 98%
CAS No.: 447-61-0
Molecular Formula: C8H5F3O
Formula Weight: 174.12
Flash Point: 61°(141°F)
Refractive Index: 1.4660
Odor: Almond-like
Melting Point: -40°
Boiling Point: 70-71°/16mm
Beilstein Reference: 2045512
MDL No.: MFCD00003337
Formula:
C₈H₅F₃O Boiling Pt: 70…71 °C/16 mm Melting Pt: –40 °C Density: 1.322 Flash Pt: 61 °C(141 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003337 CAS Number: 447-61-0 EINECS: 207-185-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...