Methyl phenylpropiolate 97%
Supplier: Thermo Scientific Chemicals
B24587-06
B24587-14
B24587-22
CAAAB24587-06EA
102.7
CAD
CAAAB24587-06
CAAAB24587-14
CAAAB24587-22
Methyl phenylpropiolate 97%
Methyl phenylpropiolate
Purity: 97%
CAS No.: 4891-38-7
Molecular Formula: C10H8O2
Formula Weight: 160.17
Flash Point:
Formula:
C₁₀H₈O₂ Boiling Pt: 109…110 °C/2 mm Melting Pt: 26 °C Density: 1.083 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041685 CAS Number: 4891-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...