4-Amino-m-toluic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24579-06
B24579-14
CAAAB24579-03EA
462.58
CAD
CAAAB24579-03
CAAAB24579-06
CAAAB24579-14
4-Amino-m-toluic acid 98%
4-Amino-m-toluic acid
Purity: 98%
CAS No.: 2486-70-6
Molecular Formula: C8H9NO2
Formula Weight: 151.16
Melting Point: 169-171°
Beilstein Reference: 2802615
Formula:
C₈H₉NO₂ Boiling Pt: 340 °C (1013 hPa) Melting Pt: 169…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007736 CAS Number: 2486-70-6 EINECS: 219-629-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...