2,5-Dimethoxyphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24571-03
B24571-06
B24571-14
CAAAB24571-03EA
21.8
CAD
CAAAB24571-03
CAAAB24571-06
CAAAB24571-14
2,5-Dimethoxyphenylboronic acid 98%
2,5-Dimethoxyphenylboronic acid
Purity: 98%
CAS No.: 107099-99-0
Molecular Formula: C8H11BO4
Formula Weight: 181.98
Melting Point: 92-94°
Beilstein Reference: 2969872
Formula:
C₈H₁₁BO₄ Melting Pt: 92…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318181 CAS Number: 107099-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...