3-Amino-2-naphthol 97%

Supplier: Thermo Scientific Chemicals
Warning

B24570-03 B24570-06 B24570-14
CAAAB24570-03EA 120.08 CAD
CAAAB24570-03 CAAAB24570-06 CAAAB24570-14
3-Amino-2-naphthol 97%
3-Amino-2-naphthol

Purity: 97% CAS No.: 5417-63-0 Molecular Formula: C10H9NO Formula Weight: 159.19 Odor: Odorless Melting Point: 232-238° MDL No.: MFCD00004113

Formula: C₁₀H₉NO
Melting Pt: 232…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00004113
CAS Number: 5417-63-0
EINECS: 226-519-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR