2-(Bromomethyl)naphthalene 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24568-06
B24568-14
B24568-22
CAAAAB24568-06EA
91.48
CAD
CAAAAB24568-06
CAAAAB24568-14
CAAAAB24568-22
2-(Bromomethyl)naphthalene 96%
2-(Bromomethyl)naphthalene
Purity: 96%
CAS No.: 939-26-4
Molecular Formula: C11H9Br
Formula Weight: 221.10
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₁H₉Br Boiling Pt: 213 °C/100 mm Melting Pt: 51…55 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004123 CAS Number: 939-26-4 EINECS: 213-359-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...