(Benzyl)triphenylphosphonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzyltriphenylphosphonium bromide
B24567-09
B24567-18
B24567-30
CAAAAB24567-09EA
42.82
CAD
CAAAAB24567-09
CAAAAB24567-18
CAAAAB24567-30
(Benzyl)triphenylphosphonium bromide 98%
(Benzyl)triphenylphosphonium bromide
Purity: 98%
CAS No.: 1449-46-3
Molecular Formula: C6H5CH2P(C6H5)3Br
Formula Weight: 433.34
Melting Point: 293-298°
Beilstein Reference: 3599867
UN No.: 3464
MDL No.: MFCD00031707
Formula:
C₂₅H₂₂BrP MW: 433.33 g/mol Melting Pt: 293…298 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031707 CAS Number: 1449-46-3 EINECS: 215-908-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...