Tetrabromophthalic anhydride 98%

Supplier: Thermo Scientific Chemicals
Warning

B24550-22 B24550-36
CAAAB24550-22EA 76 CAD
CAAAB24550-22 CAAAB24550-36
Tetrabromophthalic anhydride 98%
Tetrabromophthalic anhydride

Purity: 98% CAS No.: 632-79-1 Molecular Formula: C8Br4O3 Formula Weight: 463.70 Odor: Odorless Melting Point: 274-280° Beilstein Reference: 18908 MDL No.: MFCD00005919

Formula: C₈Br₄O₃
MW: 463.7 g/mol
Boiling Pt: 477 °C (1013 hPa)
Melting Pt: 274…280 °C
Density: 2.87
Storage Temperature: Ambient
MDL Number: MFCD00005919
CAS Number: 632-79-1
EINECS: 211-185-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR