1-Ethyl-2-pyrrolidone 98%

Supplier: Thermo Scientific Chemicals
Danger

B24548-14 B24548-22 B24548-36
CAAAB24548-14EA 66.36 CAD
CAAAB24548-14 CAAAB24548-22 CAAAB24548-36
1-Ethyl-2-pyrrolidone 98%
1-Ethyl-2-pyrrolidone

Purity: 98% CAS No.: 2687-91-4 Molecular Formula: C6H11NO Formula Weight: 113.16 Flash Point: 76°(169°F) Refractive Index: 1.4650 Boiling Point: 96-98°/20mm MDL No.: MFCD00003199

Formula: C₆H₁₁NO
Boiling Pt: 96…98 °C/20 mm
Density: 0.992
Flash Pt: 76 °C(169 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003199
CAS Number: 2687-91-4
EINECS: 220-250-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR