Isoeugenol cis- and trans-mixture 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24541-14
B24541-22
B24541-36
CAAAB24541-14EA
38.39
CAD
CAAAB24541-14
CAAAB24541-22
CAAAB24541-36
Isoeugenol cis- and trans-mixture 98+%
Isoeugenol
Purity: 98+%
CAS No.: 97-54-1
Molecular Formula: C10H12O2
Formula Weight: 164.21
Flash Point: 134°(273°F)
Refractive Index: 1.5770
Melting Point: -10°
Boiling Point: 266-268°
Beilstein Reference: 1909602
Merck Reference: 14,5171
MDL No.: MFCD00009285
Formula:
C₁₀H₁₂O₂ Boiling Pt: 266…268 °C Melting Pt: –10 °C Density: 1.080 Flash Pt: 134 °C(273 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009285 CAS Number: 97-54-1 EINECS: 202-590-7 Merck Index: 14,5171 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...