Eosin Y (yellowish)

Supplier: Thermo Scientific Chemicals
Warning

B24535-18 B24535-30
CAAAB24535-18EA 69.84 CAD
CAAAB24535-18 CAAAB24535-30
Eosin Y (yellowish)
Eosin Y (yellowish)

Fluorescent stain for cytoplasm and collagen


CAS No.: 17372-87-1 Molecular Formula: C20H6Br4Na2O5 Formula Weight: 691.86 Odor: Odorless Solubility: Soluble in water, fairly soluble in alcohol, insoluble in ether Beilstein Reference: 3586809 Merck Reference: 14,3603 MDL No.: MFCD00133309

Formula: C₂₀H₆Br₄Na₂O₅
Melting Pt: 295…296 °C
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
EINECS: 241-409-6
Merck Index: 14,3603

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR