3-Methylbutanone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Isopropyl Methyl Ketone, 3-Methyl-2-butanone
B24527-AK
B24527-AU
CAAAB24527-AKEA
85.95
CAD
CAAAB24527-AK
CAAAB24527-AU
3-Methylbutanone 98%
3-Methylbutanone
Purity: 98%
CAS No.: 563-80-4
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: -1°(30°F)
Refractive Index: 1.3880
Odor: Fruit-like
Melting Point: -92°
Boiling Point: 94-95°
Beilstein Reference: 1071238
Merck Reference: 14,6090
UN No.: 2397
MDL No.: MFCD00008919
Formula:
C₅H₁₀O Boiling Pt: 94…95 °C Melting Pt: –92 °C Density: 0.805 Flash Pt: –1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008919 CAS Number: 563-80-4 EINECS: 209-264-3 UN: 2397 ADR: 3,II Merck Index: 14,6090 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...