Methyl benzenesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger

B24515-14 B24515-22 B24515-36
CAAAB24515-14EA 75.52 CAD
CAAAB24515-14 CAAAB24515-22 CAAAB24515-36
Methyl benzenesulfonate 98%
Methyl benzenesulphonate

Purity: 98% CAS No.: 80-18-2 Molecular Formula: C7H8O3S Formula Weight: 172.20 Flash Point: 143°(289°F) Refractive Index: 1.5165 Melting Point: -4° Boiling Point: 280° Beilstein Reference: 908448 UN No.: 3265 MDL No.: MFCD00014737

Formula: C₇H₈O₃S
Boiling Pt: 280 °C
Melting Pt: –4 °C
Density: 1.269
Flash Pt: 143 °C(289 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014737
CAS Number: 80-18-2
EINECS: 201-256-8
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR