2-Amino-2-ethyl-1,3-propanediol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Amino-2-ethylpropane-1,3-diol

B24509-22 B24509-36
CAAAB24509-22EA 72.36 CAD
CAAAB24509-22 CAAAB24509-36
2-Amino-2-ethyl-1,3-propanediol 97%
2-Amino-2-ethyl-1,3-propanediol

Purity: 97% CAS No.: 115-70-8 Molecular Formula: C5H13NO2 Formula Weight: 119.16 Flash Point: 110°(230°F) Refractive Index: 1.4880 Boiling Point: 152-153°/10mm Merck Reference: 14,439 MDL No.: MFCD00004680

Formula: C₅H₁₃NO₂
MW: 119.16 g/mol
Boiling Pt: 152…153 °C/10 mm
Melting Pt: 39
Density: 1.099
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004680
CAS Number: 115-70-8
EINECS: 204-101-2
Merck Index: 14,439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR