Diglycolic anhydride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24498-09
B24498-18
CAAAB24498-09EA
92.59
CAD
CAAAB24498-09
CAAAB24498-18
Diglycolic anhydride 97%
Diglycolic anhydride
Purity: 97%
CAS No.: 4480-83-5
Molecular Formula: C4H4O4
Formula Weight: 116.07
Melting Point: 91-94°
Boiling Point: 240-241°
MDL No.: MFCD00006677
Formula:
C₄H₄O₄ Boiling Pt: 240…241 °C Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006677 CAS Number: 4480-83-5 EINECS: 224-761-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...