1,2-Propylenediamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,2-Diaminopropane
B24489-AE
B24489-AP
B24489-AU
CAAAAB24489-AEEA
40.76
CAD
CAAAAB24489-AE
CAAAAB24489-AP
CAAAB24489-AU
1,2-Propylenediamine 99%
1,2-Propylenediamine
Purity: 99%
CAS No.: 78-90-0
Molecular Formula: C3H10N2
Formula Weight: 74.13
Flash Point: 34°(93°F)
Refractive Index: 1.4460
Odor: Characteristic
Melting Point: ca -37°
Boiling Point: 119-120°
Beilstein Reference: 605274
Merck Reference: 14,7852
UN No.: 2258
MDL No.: MFCD00008089
Formula:
C₃H₁₀N₂ Boiling Pt: 119…120 °C Melting Pt: ∼ 37 °C Density: 0.869 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008089 CAS Number: 78-90-0 EINECS: 201-155-9 UN: 2258 ADR: 8,II Merck Index: 14,7852 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...