(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
(S)-(+)-1,2-O-Isopropylidenglycerol, (S)-(+)-2,2-Dimethyl-1,3-dioxolane-4-methanol, (S)-(+)-1,2-Isopropylideneglycerol, (+)-Solketal, (S)-(+)-2,3-O-Isopropylideneglycerol
Purity: 98%
CAS No.: 22323-82-6
Molecular Formula: C6H12O3
Formula Weight: 132.18
Storage Temperature: 2°C to 8°C
Flash Point: 80°(176°F)
Refractive Index: 1.4350
Boiling Point: 82-83°/14mm
Beilstein Reference: 80118
Merck Reference: 14,5213
Optical Rotation: +11.5° (c=5 in methanol)
MDL No.: MFCD00063239
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 82…83 °C/14 mm Density: 1.066 Flash Pt: 80 °C(176 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00063239 CAS Number: 22323-82-6 EINECS: 244-910-8 Merck Index: 14,5213 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...