Isobutyryl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

B24472-22 B24472-36
CAAAB24472-22EA 63.52 CAD
CAAAB24472-22 CAAAB24472-36
Isobutyryl chloride 98%
Isobutyryl chloride

Purity: 98% CAS No.: 79-30-1 Molecular Formula: (CH3)2CHCOCl Formula Weight: 106.55 Flash Point: 8°(46°F) Refractive Index: 1.4070 Odor: Unpleasant Melting Point: ca -90° Boiling Point: 91-93° Beilstein Reference: 605393 UN No.: 2395 MDL No.: MFCD00000717

Formula: C₄H₇ClO
MW: 106.55 g/mol
Boiling Pt: 91…93 °C
Melting Pt: ca …90 °C
Density: 1.017
Flash Pt: 8 °C(46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000717
CAS Number: 79-30-1
EINECS: 201-194-1
UN: 2395
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR