Isopropyl salicylate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isopropyl 2-hydroxybenzoate

B24471-14 B24471-22
CAAAB24471-14EA 109.18 CAD
CAAAB24471-14 CAAAB24471-22
Isopropyl salicylate 99%
Isopropyl salicylate

Purity: 99% CAS No.: 607-85-2 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 93°(199°F) Boiling Point: 122°/18mm MDL No.: MFCD00035703

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 122
Density: 1.09
Flash Pt: >93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00035703
CAS Number: 607-85-2
EINECS: 210-143-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR