4'-Bromo-3'-nitroacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Bromo-3-nitrophenyl)ethan-1-one, 1-(4-Bromo-3-nitrophenyl)ethanone

B24469-06 B24469-14
CAAAB24469-06EA 113.76 CAD
CAAAB24469-06 CAAAB24469-14
4'-Bromo-3'-nitroacetophenone 99%
4'-Bromo-3'-nitroacetophenone

Purity: 99% CAS No.: 18640-58-9 Molecular Formula: C8H6BrNO3 Formula Weight: 244.05 Melting Point: 117-120° Beilstein Reference: 2050098

Formula: C₈H₆BrNO₃
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00016985
CAS Number: 18640-58-9
EINECS: 242-469-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR