2,6-Dichlorotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,3-Dichloro-2-methylbenzene
B24463-14
B24463-22
B24463-36
CAAAB24463-14EA
42.82
CAD
CAAAB24463-14
CAAAB24463-22
CAAAB24463-36
2,6-Dichlorotoluene 99%
2,6-Dichlorotoluene
Purity: 99%
CAS No.: 118-69-4
Molecular Formula: C7H6Cl2
Formula Weight: 161.03
Flash Point: 82°(179°F)
Refractive Index: 1.5510
Melting Point: 2°
Boiling Point: 196-203°
Beilstein Reference: 1907194
UN No.: 3082
MDL No.: MFCD00000576
Formula:
C₇H₆Cl₂ Boiling Pt: 196…203 °C Melting Pt: 2 °C Density: 1.254 Flash Pt: 82 °C (179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000576 CAS Number: 118-69-4 EINECS: 204-269-7 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...