m-Toluamide 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Methylbenzamide
B24442-14
B24442-22
CAAAB24442-14EA
72.21
CAD
CAAAB24442-14
CAAAB24442-22
m-Toluamide 99%
m-Toluamide
Purity: 99%
CAS No.: 618-47-3
Molecular Formula: C8H9NO
Formula Weight: 135.17
Melting Point: 93-96°
Beilstein Reference: 1857374
Formula:
C₈H₉NO MW: 135.17 g/mol Melting Pt: 93…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007990 CAS Number: 618-47-3 EINECS: 210-553-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...