1,2-Diiodobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24425-03
B24425-06
B24425-14
CAAAB24425-03EA
60.83
CAD
CAAAB24425-03
CAAAB24425-06
CAAAB24425-14
1,2-Diiodobenzene 98%
1,2-Diiodobenzene
Purity: 98%
CAS No.: 615-42-9
Molecular Formula: C6H4I2
Formula Weight: 329.91
Flash Point:
Formula:
C₆H₄I₂ Boiling Pt: 282 °C Density: 2.524 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001038 CAS Number: 615-42-9 EINECS: 210-425-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...