Sodium hydroxide 97%, pearl

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Caustic soda

B24414-0E B24414-36
CAAAAB24414-0EEA 135.56 CAD
CAAAAB24414-0E CAAAAB24414-22 CAAAAB24414-36
Sodium hydroxide 97%, pearl
Sodium hydroxide

Purity: 97% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Odor: Odorless pH: 14 Melting Point: 318° Boiling Point: 1390° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR