3-Fluoro-4-(trifluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Fluoro-4-trifluoromethylaniline, 4-Amino-alpha,alpha,alpha,2-tetrafluorotoluene, α,α,α,3-Tetrafluoro-p-toluidine, 4-Amino-2-fluorobenzotrifluoride, 4-Amino-α,α,α,2-tetrafluorotoluene

B24411-03 B24411-06
CAAAAB24411-03EA 51.51 CAD
CAAAAB24411-03 CAAAAB24411-06
3-Fluoro-4-(trifluoromethyl)aniline 97%
3-Fluoro-4-(trifluoromethyl)aniline

Purity: 97% CAS No.: 69411-68-3 Molecular Formula: C7H5F4N Formula Weight: 179.12 Melting Point: 54-58° UN No.: 2811 MDL No.: MFCD00190120

Formula: C₇H₅F₄N
MW: 179.12 g/mol
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00190120
CAS Number: 69411-68-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR