(±)-2-Methylpiperazine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24405-14
B24405-22
CAAAAB24405-14EA
70.63
CAD
CAAAAB24405-14
CAAAAB24405-22
(±)-2-Methylpiperazine 98%
(±)-2-Methylpiperazine
Purity: 98%
CAS No.: 109-07-9
Molecular Formula: C5H12N2
Formula Weight: 100.17
Flash Point: 65°(149°F)
Odor: Amine-like
Boiling Point: 155-156°
Beilstein Reference: 383566
UN No.: 2925
MDL No.: MFCD00005954
Formula:
C₅H₁₂N₂ Boiling Pt: 155…156 °C Melting Pt: 62…66 °C Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005954 CAS Number: 109-07-9 EINECS: 203-644-2 UN: 2925 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...