(2-Methoxyphenyl)acetonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methoxybenzyl cyanide
B24400-0B
B24400-18
B24400-30
CAAAB24400-0BEA
753.66
CAD
CAAAB24400-0B
CAAAB24400-18
CAAAB24400-30
(2-Methoxyphenyl)acetonitrile 98%
(2-Methoxyphenyl)acetonitrile
Purity: 98%
CAS No.: 7035-03-2
Molecular Formula: C9H9NO
Formula Weight: 147.18
Flash Point: 129°(264°F)
Melting Point: 66-70°
Boiling Point: 141-143°/15mm
Beilstein Reference: 2087748
UN No.: 3439
MDL No.: MFCD00001902
Formula:
C₉H₉NO Boiling Pt: 141…143 °C/15 mm Melting Pt: 66…70 °C Flash Pt: 129 °C (264 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001902 CAS Number: 7035-03-2 EINECS: 230-314-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...