2,2-Dimethylsuccinic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2-Dimethyl-1,4-butanedioic acid, α,α-Dimethylsuccinic acid

B24394-06 B24394-14
CAAAB24394-03EA 32.72 CAD
CAAAB24394-03 CAAAB24394-06 CAAAB24394-14
2,2-Dimethylsuccinic acid 99%
2,2-Dimethylsuccinic acid

Purity: 99% CAS No.: 597-43-3 Molecular Formula: C6H10O4 Formula Weight: 146.14 Melting Point: 139-142° Beilstein Reference: 1764476

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 296
Melting Pt: 143
Storage Temperature: Ambient
MDL Number: MFCD00004196
CAS Number: 597-43-3
EINECS: 209-899-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR