3,4-Diaminotoluene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Diamino-4-methylbenzene, Toluene-3,4-diamine

B24378-22 B24378-36
CAAAB24378-22EA 60.83 CAD
CAAAB24378-22 CAAAB24378-36
3,4-Diaminotoluene 97%
3,4-Diaminotoluene

Purity: 97% CAS No.: 496-72-0 Molecular Formula: C7H10N2 Formula Weight: 122.17 Melting Point: 86-90° Boiling Point: 265° MDL No.: MFCD00007728

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 265 °C
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007728
CAS Number: 496-72-0
EINECS: 207-826-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR