2,4-Dimethoxyphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B24374-03 B24374-06 B24374-14
CAAAB24374-03EA 79 CAD
CAAAB24374-03 CAAAB24374-06 CAAAB24374-14
2,4-Dimethoxyphenylboronic acid 98%
2,4-Dimethoxyphenylboronic acid

Purity: 98% CAS No.: 133730-34-4 Molecular Formula: C8H11BO4 Formula Weight: 181.98 Melting Point: 115-120° Beilstein Reference: 4744213

Formula: C₈H₁₁BO₄
MW: 181.98
Melting Pt: 115…120 °C
Storage Temperature: Ambient
MDL Number: MFCD01074590
CAS Number: 133730-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR