4-Bromo-2-fluorobenzenesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24352-06
B24352-14
CAAAB24352-03EA
116.48
CAD
CAAAB24352-03
CAAAB24352-06
CAAAB24352-14
4-Bromo-2-fluorobenzenesulfonyl chloride 98%
4-Bromo-2-fluorobenzenesulphonyl chloride
Purity: 98%
CAS No.: 216159-03-4
Molecular Formula: C6H3BrClFO2S
Formula Weight: 273.51
Melting Point: 63-66°
Boiling Point: 84°/0.5mm
UN No.: 3261
MDL No.: MFCD01090993
Formula:
C₆H₃BrClFO₂S MW: 273.51 Boiling Pt: 84 °C/0.5 mm Melting Pt: 63…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD01090993 CAS Number: 216159-03-4 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...