4-Bromobenzamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24348-04
B24348-09
B24348-18
CAAAB24348-04EA
60.04
CAD
CAAAB24348-04
CAAAB24348-09
CAAAB24348-18
4-Bromobenzamide 97%
4-Bromobenzamide
Purity: 97%
CAS No.: 698-67-9
Molecular Formula: C7H6BrNO
Formula Weight: 200.03
Melting Point: 190-193°
MDL No.: MFCD00007991
Formula:
C₇H₆BrNO Boiling Pt: 328 °C (1013 hPa) Melting Pt: 190…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007991 CAS Number: 698-67-9 EINECS: 211-817-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...