2-(Trifluoromethyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Trifluoromethyl)phenylboronic acid, (2-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethyl)phenyl)borane, 2-(Trifluoromethyl)benzeneboronic acid

B24343-03 B24343-06 B24343-14
CAAAB24343-03EA 65.25 CAD
CAAAB24343-03 CAAAB24343-06 CAAAB24343-14
2-(Trifluoromethyl)phenylboronic acid 97%
2-(Trifluoromethyl)phenylboronic acid

Purity: 97% CAS No.: 1423-27-4 Molecular Formula: C7H6BF3O2 Formula Weight: 189.93 Melting Point: 108-110° Beilstein Reference: 2974465

Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00236059
CAS Number: 1423-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR