1H,1H-Pentafluoropropyl methacrylate 97% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H,1H-Pentafluoropropyl methacrylate, 2,2,3,3,3-Pentafluoropropyl Methacrylate, Methacrylic acid 2,2,2,3,3-pentafluoropropyl ester, Methacrylic acid 2,2,3,3,3-Pentafluoropropyl ester, 1H,1H-Perfluoropropyl methacrylate
B24340-06
B24340-14
CAAAB24340-06EA
125.93
CAD
CAAAB24340-06
CAAAB24340-14
1H,1H-Pentafluoropropyl methacrylate 97% stabilised
1H,1H-Pentafluoropropyl methacrylate
Purity: 97%
CAS No.: 45115-53-5
Molecular Formula: C7H7F5O2
Formula Weight: 218.12
Storage Temperature: 2°C to 8°C
Flash Point: 29°(84°F)
Refractive Index: 1.3480
Boiling Point: 54-55°/100mm
UN No.: 3272
MDL No.: MFCD00039256
Formula:
C₇H₇F₅O₂ MW: 218.12 Boiling Pt: 54…55 °C/100 mm Density: 1.279 Flash Pt: 29 °C (84 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039256 CAS Number: 45115-53-5 EINECS: 256-191-8 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...