4-Amino-3-hydroxy-1-naphthalenesulfonic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24339-14
B24339-22
CAAAAB24339-14EA
106.02
CAD
CAAAAB24339-14
CAAAAB24339-22
4-Amino-3-hydroxy-1-naphthalenesulfonic acid 99%
4-Amino-3-hydroxy-1-naphthalenesulphonic acid
Purity: 99%
CAS No.: 116-63-2
Molecular Formula: C10H9NO4S
Formula Weight: 239.25
Melting Point: ca 295° dec.
Beilstein Reference: 2697469
Merck Reference: 14,454
MDL No.: MFCD00004019
Formula:
C₁₀H₉NO₄S Boiling Pt: 466 °C (1013 hPa) Melting Pt: ca 295 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004019 CAS Number: 116-63-2 EINECS: 204-147-3 Merck Index: 14,454 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...