5-Amino-2,2-difluoro-1,3-benzodioxole 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24336-03
B24336-06
CAAAAB24336-03EA
97.17
CAD
CAAAAB24336-03
CAAAAB24336-06
5-Amino-2,2-difluoro-1,3-benzodioxole 97+%
5-Amino-2,2-difluoro-1,3-benzodioxole
Purity: 97+%
CAS No.: 1544-85-0
Molecular Formula: C7H5F2NO2
Formula Weight: 173.12
Refractive Index: 1.4990
Odor: Recognizable
Boiling Point: 95-97°/12mm
Beilstein Reference: 1343593
UN No.: 2810
MDL No.: MFCD00190144
Formula:
C₇H₅F₂NO₂ Boiling Pt: 95…97 °C/12 mm Storage Temperature: Ambient |
MDL Number:
MFCD00190144 CAS Number: 1544-85-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...