4-Cyanobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24333-06
B24333-14
B24333-22
CAAAB24333-06EA
90.06
CAD
CAAAB24333-06
CAAAB24333-14
CAAAB24333-22
4-Cyanobenzoyl chloride 98%
4-Cyanobenzoyl chloride
Purity: 98%
CAS No.: 6068-72-0
Molecular Formula: C8H4ClNO
Formula Weight: 165.58
Odor: Irritating
Melting Point: 68-70°
Beilstein Reference: 386729
UN No.: 3261
MDL No.: MFCD00001822
Formula:
C₈H₄ClNO Boiling Pt: 150 - 151 °C (25 -rr) Melting Pt: 68…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001822 CAS Number: 6068-72-0 EINECS: 228-005-5 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...