Diaminomaleonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

B24328-14 B24328-22
CAAAB24328-14EA 46.61 CAD
CAAAB24328-14 CAAAB24328-22
Diaminomaleonitrile 98%
Diaminomaleonitrile

Purity: 98% CAS No.: 1187-42-4 Molecular Formula: C4H4N4 Formula Weight: 108.10 Melting Point: 178-182° dec. Beilstein Reference: 878333 UN No.: 3439 MDL No.: MFCD00001870

Formula: C₄H₄N₄
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 178…182 °C (dec.)
Density: 1.360
Storage Temperature: Ambient
MDL Number: MFCD00001870
CAS Number: 1187-42-4
EINECS: 214-697-6
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR