1,1,1-Tris(hydroxymethyl)ethane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Hydroxymethyl)-2-methyl-1,3-propanediol, Ethylidynetrimethanol
B24324-0E
B24324-36
CAAAB24324-0EEA
178.54
CAD
CAAAB24324-0E
CAAAB24324-36
1,1,1-Tris(hydroxymethyl)ethane 97%
1,1,1-Tris(hydroxymethyl)ethane
Purity: 97%
CAS No.: 77-85-0
Molecular Formula: C5H12O3
Formula Weight: 120.15
Flash Point: 160°(320°F)
Melting Point: ca 190°
Boiling Point: 137°/15mm
Beilstein Reference: 1304452
MDL No.: MFCD00004687
Formula:
C₅H₁₂O₃ MW: 120.15 g/mol Boiling Pt: 287 Melting Pt: ∼ 190 °C Flash Pt: 160 °C (320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004687 CAS Number: 77-85-0 EINECS: 201-063-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...