3-Chloropyridine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24316-09
B24316-18
CAAAB24316-09EA
51.35
CAD
CAAAB24316-09
CAAAB24316-18
3-Chloropyridine 99%
3-Chloropyridine
Purity: 99%
CAS No.: 626-60-8
Molecular Formula: C5H4ClN
Formula Weight: 113.55
Flash Point: 53°(127°F)
Refractive Index: 1.5330
Boiling Point: 147-149°
Beilstein Reference: 105744
UN No.: 1993
MDL No.: MFCD00006375
Formula:
C₅H₄ClN MW: 113.55 g/mol Boiling Pt: 148 Melting Pt: -0.5 Density: 1.205 Flash Pt: 53 °C(127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006375 CAS Number: 626-60-8 EINECS: 210-955-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...